4-Methoxy-3-nitrophenylboronsäure
Lieferant: COMBI-BLOCKS
BB-5076-5G
BB-5076-1G
COBBBB-5076-5GEA
0
EUR
COBBBB-5076-5G
COBBBB-5076-1G
4-Methoxy-3-nitrophenylboronsäure
4-Methoxy-3-nitrophenylboronsäure
Formel:
C₇H₈BNO₅ |
MDL:
MFCD03092922 CAS-Nummer: 827614-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...