3-Brom-2-methoxy-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-2-methoxy-4-pyridinylboronsäure, 3-Brom-2-methoxypyridin-4-boronsäure

BB-5137-1G
COBBBB-5137-1GEA 303 EUR
COBBBB-5137-1G
3-Brom-2-methoxy-4-pyridinboronsäure
3-Bromo-2-methoxy-4-pyridineboronic acid
Formel: C₆H₇BBrNO₃
MDL: MFCD09972108
CAS-Nummer: 1072946-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR