4-(Methylsulfonyl)-2-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5177-5G BB-5177-1G
COBBBB-5177-5GEA 0 EUR
COBBBB-5177-5G COBBBB-5177-1G
4-(Methylsulfonyl)-2-(trifluormethyl)phenylboronsäure
4-(Methylsulfonyl)-2-(trifluormethyl)phenylboronsäure
Formel: C₈H₈BF₃O₄S
MDL: MFCD10699623
CAS-Nummer: 1072946-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR