2-Fluor-5-nitrophenylboronsäure
Lieferant: COMBI-BLOCKS
BB-5186-1G
COBBBB-5186-1GEA
0
EUR
COBBBB-5186-1G
2-Fluor-5-nitrophenylboronsäure
2-Fluor-5-nitrophenylboronsäure
Formel:
C₆H₅BFNO₄ |
MDL:
MFCD03092929 CAS-Nummer: 819849-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...