2-Chlor-4-methoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-5189-1G
COBBBB-5189-1GEA 0 EUR
COBBBB-5189-1G
2-Chlor-4-methoxy-3-pyridinboronsäure
2-Chloro-4-methoxy-3-pyridineboronic acid
Formel: C₆H₇BClNO₃
MDL: MFCD09972119
CAS-Nummer: 1072946-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR