4-Methoxy-3-pyridinboronsäure Hydrochlorid
Lieferant: COMBI-BLOCKS
BB-5210-1G
BB-5210-5G
COBBBB-5210-1GEA
0
EUR
COBBBB-5210-1G
COBBBB-5210-5G
4-Methoxy-3-pyridinboronsäure Hydrochlorid
4-Methoxy-3-pyridinboronsäure Hydrochlorid
Formel:
C₆H₉BClNO₃ |
MDL:
MFCD04113800 CAS-Nummer: 874959-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...