4-Benzyloxy-3,5-dichlorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5222-1G
COBBBB-5222-1GEA 0 EUR
COBBBB-5222-1G
4-Benzyloxy-3,5-dichlorphenylboronsäure
4-Benzyloxy-3,5-dichlorphenylboronsäure
Formel: C₁₃H₁₁BCl₂O₃
MDL: MFCD04039002
CAS-Nummer: 1003298-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR