5-Chlor-2-methyl-4-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5223-100M
COBBBB-5223-100MEA 0 EUR
COBBBB-5223-100M
5-Chlor-2-methyl-4-(trifluormethyl)phenylboronsäure
5-Chlor-2-methyl-4-(trifluormethyl)phenylboronsäure
Formel: C₈H₇BClF₃O₂
MDL: MFCD11053801
CAS-Nummer: 1072946-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR