3,4,5-Trichlor-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5224-100M
COBBBB-5224-100MEA 864 EUR
COBBBB-5224-100M
3,4,5-Trichlor-2-methylphenylboronsäure
3,4,5-Trichlor-2-methylphenylboronsäure
Formel: C₇H₆BCl₃O₂
MDL: MFCD11053802
CAS-Nummer: 1072946-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR