2,6-Dichlor-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5227-5G BB-5227-1G
COBBBB-5227-5GEA 0 EUR
COBBBB-5227-5G COBBBB-5227-1G
2,6-Dichlor-3-nitrophenylboronsäure
2,6-Dichlor-3-nitrophenylboronsäure
Formel: C₆H₄BCl₂NO₄
MDL: MFCD10699640
CAS-Nummer: 1072946-37-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR