2,3,4-Trichlor-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5228-1G
COBBBB-5228-1GEA 692 EUR
COBBBB-5228-1G
2,3,4-Trichlor-5-nitrophenylboronsäure
2,3,4-Trichlor-5-nitrophenylboronsäure
Formel: C₆H₃BCl₃NO₄
MDL: MFCD11053804
CAS-Nummer: 1072946-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR