2-Fluor-3-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Fluoro-3-methylphenyl)boronic acid

BB-5230-5G BB-5230-25G BB-5230-1G
COBBBB-5230-5GEA 146 EUR
COBBBB-5230-5G COBBBB-5230-25G COBBBB-5230-1G
2-Fluor-3-methylphenylboronsäure
2-Fluor-3-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153.95 g/mol
MDL: MFCD06409950
CAS-Nummer: 762287-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR