6-Chlor-2-methoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Chloro-2-methoxypyridin-3-yl)boronic acid

BB-5247-1G
COBBBB-5247-1GEA 98.3 EUR
COBBBB-5247-1G
6-Chlor-2-methoxy-3-pyridinboronsäure
6-Chloro-2-methoxy-3-pyridineboronic acid
Formel: C₆H₇BClNO₃
Molecular Weight: 187,39 g/mol
MDL: MFCD11053817
CAS-Nummer: 1072946-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR