4-Methoxy-1-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Methoxynaphthalen-1-yl)boronic acid, 4-Methoxynaphthalene-1-boronic acid

BB-5250-1G
COBBBB-5250-1GEA 486 EUR
COBBBB-5250-1G
4-Methoxy-1-naphthalinboronsäure
4-Methoxy-1-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₃
Molecular Weight: 202,0142 g/mol
MDL: MFCD03452757
CAS-Nummer: 219834-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR