2-(Brommethyl)-3,4,5-trichlorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5258-100M
COBBBB-5258-100MEA 864 EUR
COBBBB-5258-100M
2-(Brommethyl)-3,4,5-trichlorphenylboronsäure
2-(Brommethyl)-3,4,5-trichlorphenylboronsäure
Formel: C₇H₅BBrCl₃O₂
MDL: MFCD11053838
CAS-Nummer: 1072946-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR