2-(Methoxycarbonyl)-3-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5282-1G BB-5282-250M
COBBBB-5282-1GEA 864 EUR
COBBBB-5282-1G COBBBB-5282-250M
2-(Methoxycarbonyl)-3-(trifluormethyl)phenylboronsäure
2-(Methoxycarbonyl)-3-(trifluormethyl)phenylboronsäure
Formel: C₉H₈BF₃O₄
MDL: MFCD12025963
CAS-Nummer: 1150114-32-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR