1-Boc-5-chlor-2-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1-(tert-Butoxycarbonyl)-5-chloro-1H-indol-2-yl)boronic acid

BB-5312-1G
COBBBB-5312-1GEA 175 EUR
COBBBB-5312-1G
1-Boc-5-chlor-2-indolboronsäure
1-Boc-5-chlor-2-indolboronsäure
Formel: C₁₃H₁₅BClNO₄
Molecular Weight: 295,53 g/mol
MDL: MFCD04039005
CAS-Nummer: 475102-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR