6-Fluorchinolin-8-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Fluoroquinolin-8-yl)boronic acid

BB-5324-1G
COBBBB-5324-1GEA 595 EUR
COBBBB-5324-1G
6-Fluorchinolin-8-boronsäure
6-Fluorchinolin-8-boronsäure
Formel: C₉H₇BFNO₂
Molecular Weight: 190,97 g/mol
MDL: MFCD11504849
CAS-Nummer: 1072951-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR