2-Hydroxy-4-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Hydroxy-4-(trifluoromethyl)phenyl)boronic acid

BB-5333-1G
COBBBB-5333-1GEA 528 EUR
COBBBB-5333-1G
2-Hydroxy-4-(trifluormethyl)phenylboronsäure
2-Hydroxy-4-(trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₃
Molecular Weight: 205,9270 g/mol
MDL: MFCD03094996
CAS-Nummer: 1072951-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR