2,6-Dichlor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-5391-1G
COBBBB-5391-1GEA 0 EUR
COBBBB-5391-1G
2,6-Dichlor-4-pyridinboronsäure
2,6-Dichloro-4-pyridineboronic acid
Formel: C₅H₄BCl₂NO₂
MDL: MFCD09751352
CAS-Nummer: 1072951-54-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR