5-(Trifluormethyl)chinolin-8-boronsäure
Lieferant: COMBI-BLOCKS
BB-5393-1G
COBBBB-5393-1GEA
1370
EUR
COBBBB-5393-1G
5-(Trifluormethyl)chinolin-8-boronsäure
5-(Trifluormethyl)chinolin-8-boronsäure
Formel:
C₁₀H₇BF₃NO₂ |
MDL:
MFCD11504856 CAS-Nummer: 1072951-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...