3-Methoxy-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Methoxy-5-methylphenyl)boronic acid

BB-5507-1G
COBBBB-5507-1GEA 595 EUR
COBBBB-5507-1G
3-Methoxy-5-methylphenylboronsäure
3-Methoxy-5-methylphenylboronsäure
Formel: C₈H₁₁BO₃
Molecular Weight: 165,9821 g/mol
MDL: MFCD09952026
CAS-Nummer: 725251-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR