5-(tert-Butyldimethylsilyloxy)-2,3-difluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5535-1G
COBBBB-5535-1GEA 623 EUR
COBBBB-5535-1G
5-(tert-Butyldimethylsilyloxy)-2,3-difluorphenylboronsäure
5-(tert-Butyldimethylsilyloxy)-2,3-difluorphenylboronsäure
Formel: C₁₂H₁₉BF₂O₃Si
MDL: MFCD11855842
CAS-Nummer: 1150114-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR