4-Chlor-3,5-dimethylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dimethyl-4-chlorophenylboronic acid, (4-Chloro-3,5-dimethylphenyl)boronic acid

BB-5541-1G
COBBBB-5541-1GEA 385 EUR
COBBBB-5541-1G
4-Chlor-3,5-dimethylphenylboronsäure
4-Chlor-3,5-dimethylphenylboronsäure
Formel: C₈H₁₀BClO₂
Molecular Weight: 184,4278 g/mol
MDL: MFCD10566596
CAS-Nummer: 1056475-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR