4-Cyan-3-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Cyano-3-(trifluoromethyl)phenyl)boronic acid

BB-5560-1G
COBBBB-5560-1GEA 214 EUR
COBBBB-5560-1G
4-Cyan-3-(trifluormethyl)phenylboronsäure
4-Cyan-3-(trifluormethyl)phenylboronsäure
Formel: C₈H₅BF₃NO₂
Molecular Weight: 214,937 g/mol
MDL: MFCD10697412
CAS-Nummer: 915299-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR