6-(Methoxycarbonyl)-2-indolboronsäure

Lieferant: COMBI-BLOCKS

BB-5577-1G
COBBBB-5577-1GEA 0 EUR
COBBBB-5577-1G
6-(Methoxycarbonyl)-2-indolboronsäure
6-(Methoxycarbonyl)-2-indolboronsäure
Formel: C₁₀H₁₀BNO₄
MDL: MFCD11855843
CAS-Nummer: 1150114-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR