2-(Trifluormethyl)-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Trifluoromethyl)pyridin-4-yl)boronic acid

BB-5671-1G
COBBBB-5671-1GEA 514 EUR
COBBBB-5671-1G
2-(Trifluormethyl)-4-pyridinboronsäure
2-(Trifluoromethyl)-4-pyridineboronic acid
Formel: C₆H₅BF₃NO₂
Molecular Weight: 190,9156 g/mol
MDL: MFCD11616522
CAS-Nummer: 1093407-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR