2-Methyl-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Methylpyrimidin-5-yl)boronic acid, 2-Methylpyrimidine-5-boronic acid

BB-5686-1G
COBBBB-5686-1GEA 345 EUR
COBBBB-5686-1G
2-Methyl-5-pyrimidinboronsäure
2-Methyl-5-pyrimidineboronic acid
Formel: C₅H₇BN₂O₂
Molecular Weight: 137,9323 g/mol
MDL: MFCD07375144
CAS-Nummer: 1034924-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR