2-(Ethoxycarbonyl)furan-3-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Ethoxycarbonyl)furan-3-yl)boronic acid

BB-5716-1G
COBBBB-5716-1GEA 1020 EUR
COBBBB-5716-1G
2-(Ethoxycarbonyl)furan-3-boronsäure
2-(Ethoxycarbonyl)furan-3-boronsäure
Formel: C₇H₉BO₅
Molecular Weight: 183,9544 g/mol
MDL: MFCD12025973
CAS-Nummer: 1150114-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR