3-(Ethoxycarbonylmethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-(2-Ethoxy-2-oxoethyl)phenyl)boronic acid
BB-5817-1G
COBBBB-5817-1GEA
356
EUR
COBBBB-5817-1G
3-(Ethoxycarbonylmethyl)phenylboronsäure
3-(Ethoxycarbonylmethyl)phenylboronsäure
Formel:
C₁₀H₁₃BO₄ Molecular Weight: 208,0200 g/mol |
MDL:
MFCD08704681 CAS-Nummer: 1256345-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...