3,4-Difluor-(5-methoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3,4-Difluoro-5-(methoxycarbonyl)phenylboronic acid
BB-5826-250M
COBBBB-5826-250MEA
1490
EUR
COBBBB-5826-250M
3,4-Difluor-(5-methoxycarbonyl)phenylboronsäure
3,4-Difluor-(5-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₇BF₂O₄ |
MDL:
MFCD12025989 CAS-Nummer: 1217500-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...