2-(Chlormethyl)-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5831-5G BB-5831-1G BB-5831-250M
COBBBB-5831-5GEA 0 EUR
COBBBB-5831-5G COBBBB-5831-1G COBBBB-5831-250M
2-(Chlormethyl)-5-nitrophenylboronsäure
2-(Chlormethyl)-5-nitrophenylboronsäure
Formel: C₇H₇BClNO₄
MDL: MFCD12025991
CAS-Nummer: 1217500-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR