4-Brom-2-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromo-2-nitrophenyl)boronic acid

BB-5834-250M
COBBBB-5834-250MEA 0 EUR
COBBBB-5834-250M
4-Brom-2-nitrophenylboronsäure
4-Brom-2-nitrophenylboronsäure
Formel: C₆H₅BBrNO₄
MDL: MFCD12025992
CAS-Nummer: 860034-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR