2-(Tetrahydropyran-2-yloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5858-1G
COBBBB-5858-1GEA 0 EUR
COBBBB-5858-1G
2-(Tetrahydropyran-2-yloxy)phenylboronsäure
2-(Tetrahydropyran-2-yloxy)phenylboronsäure
Formel: C₁₁H₁₅BO₄
MDL: MFCD06801685
CAS-Nummer: 502159-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR