4-(trans-4-Pentylcyclohexyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-5915-1G
COBBBB-5915-1GEA
38.5
EUR
COBBBB-5915-1G
4-(trans-4-Pentylcyclohexyl)phenylboronsäure
4-(trans-4-Pentylcyclohexyl)phenylboronsäure
Formel:
C₁₇H₂₇BO₂ Molecular Weight: 274,21 g/mol |
MDL:
MFCD09750938 CAS-Nummer: 143651-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...