4-(trans-4-Ethylcyclohexyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(trans-4-Ethylcyclohexyl)phenyl)boronic acid

BB-5916-1G
COBBBB-5916-1GEA 303 EUR
COBBBB-5916-1G
4-(trans-4-Ethylcyclohexyl)phenylboronsäure
4-(trans-4-Ethylcyclohexyl)phenylboronsäure
Formel: C₁₄H₂₁BO₂
Molecular Weight: 232,1263 g/mol
MDL: MFCD12546553
CAS-Nummer: 164220-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR