2-Brom-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Bromo-4-fluorophenyl)boronic acid

BB-5964-1G BB-5964-5G BB-5964-250M
COBBBB-5964-1GEA 0 EUR
COBBBB-5964-1G COBBBB-5964-5G COBBBB-5964-250M
2-Brom-4-fluorphenylboronsäure
2-Brom-4-fluorphenylboronsäure
Formel: C₆H₅BBrFO₂
MDL: MFCD13195647
CAS-Nummer: 1217501-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR