1-(2,2-Diethoxyethyl)-1H-pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-(2,2-Diethoxyethyl)pyrazole-4-boronic acid

BB-6004-1G
COBBBB-6004-1GEA 595 EUR
COBBBB-6004-1G
1-(2,2-Diethoxyethyl)-1H-pyrazol-4-boronsäure
1-(2,2-Diethoxyethyl)-1H-pyrazole-4-boronic acid
Formel: C₉H₁₇BN₂O₄
Molecular Weight: 228,05 g/mol
MDL: MFCD12756418
CAS-Nummer: 1217501-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR