1-(1-Boc-piperidino)-1H-pyrazol-4-boronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(1-(1-(tert-Butoxycarbonyl)piperidin-4-yl)-1H-pyrazol-4-yl)boronic acid
BB-6007-1G
BB-6007-250M
COBBBB-6007-1GEA
719
EUR
COBBBB-6007-1G
COBBBB-6007-250M
1-(1-Boc-piperidino)-1H-pyrazol-4-boronsäure
1-(1-Boc-piperidino)-1H-pyrazol-4-boronsäure
Formel:
C₁₃H₂₂BN₃O₄ Molecular Weight: 295.1425 g/mol |
MDL:
MFCD12913942 CAS-Nummer: 1190875-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...