5-Chlor-2-isopropoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Chloro-2-isopropoxypyridin-3-yl)boronic acid, 5-Chloro-2-isopropoxypyridine-3-boronic acid

BB-6125-1G
COBBBB-6125-1GEA 302 EUR
COBBBB-6125-1G
5-Chlor-2-isopropoxy-3-pyridinboronsäure
5-Chlor-2-isopropoxy-3-pyridinboronsäure
Formel: C₈H₁₁BClNO₃
Molecular Weight: 215,4400 g/mol
MDL: MFCD13195662
CAS-Nummer: 1217501-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR