5-Chlor-2-isobutoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-6126-1G
COBBBB-6126-1GEA 0 EUR
COBBBB-6126-1G
5-Chlor-2-isobutoxy-3-pyridinboronsäure
5-Chlor-2-isobutoxy-3-pyridinboronsäure
Formel: C₉H₁₃BClNO₃
MDL: MFCD13195663
CAS-Nummer: 1217501-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR