5-Chlor-2-propoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-6127-1G
COBBBB-6127-1GEA 451 EUR
COBBBB-6127-1G
5-Chlor-2-propoxy-3-pyridinboronsäure
5-Chloro-2-propoxy-3-pyridineboronic acid
Formel: C₈H₁₁BClNO₃
MDL: MFCD13195664
CAS-Nummer: 1217501-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR