4-Benzyloxy-3-chlor-5-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-6218-1G
COBBBB-6218-1GEA
404
EUR
COBBBB-6218-1G
4-Benzyloxy-3-chlor-5-methoxyphenylboronsäure
4-Benzyloxy-3-chlor-5-methoxyphenylboronsäure
Formel:
C₁₄H₁₄BClO₄ |
MDL:
MFCD15143463 CAS-Nummer: 1218790-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...