2-Isobutoxy-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-6366-1G
COBBBB-6366-1GEA 0 EUR
COBBBB-6366-1G
2-Isobutoxy-5-(trifluormethyl)phenylboronsäure
2-Isobutoxy-5-(trifluormethyl)phenylboronsäure
Formel: C₁₁H₁₄BF₃O₃
MDL: MFCD16036116
CAS-Nummer: 1256345-99-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR