(Pyridin-2-ylthio)methylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: ((Pyridin-2-ylthio)methyl)boronic acid

BB-6372-1G
COBBBB-6372-1GEA 1110 EUR
COBBBB-6372-1G
(Pyridin-2-ylthio)methylboronsäure
(Pyridin-2-ylthio)methylboronsäure
Formel: C₆H₈BNO₂S
Molecular Weight: 169,92 g/mol
MDL: MFCD16295062
CAS-Nummer: 1256346-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR