4-Acetyl-2-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-6442-1G BB-6442-250M
COBBBB-6442-1GEA 0 EUR
COBBBB-6442-1G COBBBB-6442-250M
4-Acetyl-2-nitrophenylboronsäure
4-Acetyl-2-nitrophenylboronsäure
Formel: C₈H₈BNO₅
MDL: MFCD16618934
CAS-Nummer: 1126895-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR