2,3,5,6-Tetramethylphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8001-1G
COBBBB-8001-1GEA 98.3 EUR
COBBBB-8001-1G
2,3,5,6-Tetramethylphenylboronsäure
2,3,5,6-Tetramethylphenylboronsäure
Formel: C₁₀H₁₅BO₂
Molecular Weight: 178.04 g/mol
MDL: MFCD03093882
CAS-Nummer: 197223-36-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR