2-(2'-Methoxybenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8003-1G
COBBBB-8003-1GEA 149 EUR
COBBBB-8003-1G
2-(2'-Methoxybenzyloxy)phenylboronsäure
2-(2'-Methoxybenzyloxy)phenylboronsäure
Formel: C₁₄H₁₅BO₄
MDL: MFCD07784390
CAS-Nummer: 871125-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR