2,3,5-Trifluorphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-8009-1G
COBBBB-8009-1GEA
98.3
EUR
COBBBB-8009-1G
2,3,5-Trifluorphenylboronsäure
2,3,5-Trifluorphenylboronsäure
Formel:
C₆H₄BF₃O₂ Molecular Weight: 175,9 g/mol |
MDL:
MFCD01863166 CAS-Nummer: 247564-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...