3-Benzo[b]thiophenboronsäure
Lieferant: COMBI-BLOCKS
BB-8012-1G
COBBBB-8012-1GEA
0
EUR
COBBBB-8012-1G
3-Benzo[b]thiophenboronsäure
3-Benzo[b]thiophenboronsäure
Formel:
C₈H₇BO₂S |
MDL:
MFCD01114669 CAS-Nummer: 113893-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...